siRNA Details
| Gene Symbol: Ufl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126279.1 | NP_001119751.1 | 2 | 190 | ||
| XM_006237959.2 | XP_006238021.1 | 2 | 594 | |||
| GenBank mRNA | BC166498.1 | - | 2 | 190 | 7114599 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ufl1| Gene Symbol: Ufl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001126279.1 | NP_001119751.1 | 2 | 190 | ||
| XM_006237959.2 | XP_006238021.1 | 2 | 594 | |||
| GenBank mRNA | BC166498.1 | - | 2 | 190 | 7114599 | |