siRNA Details
| Gene Symbol: Gja10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173508.1 | NP_001166979.1 | 1 | 878 | ||
| XM_008763619.1 | XP_008761841.1 | 2 | 1257 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gja10| Gene Symbol: Gja10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001173508.1 | NP_001166979.1 | 1 | 878 | ||
| XM_008763619.1 | XP_008761841.1 | 2 | 1257 | |||