siRNA Details
| Gene Symbol: Ubap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107928.1 | NP_001101398.1 | 8 | 737 | ||
| XM_006238051.2 | XP_006238113.1 | 8 | 669 | |||
| XM_006238052.2 | XP_006238114.1 | 7 | 633 | |||
| XM_006238053.2 | XP_006238115.1 | 8 | 669 | |||
| XM_006238054.2 | XP_006238116.1 | 7 | 633 | |||
| XM_008763625.1 | XP_008761847.1 | 7 | 1175 | |||

Ubap2
Genomic Map