siRNA Details
| Gene Symbol: Hsdl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025697.1 | NP_001020868.1 | 4, 5 | 506 | ||
| XM_008763735.1 | XP_008761957.1 | 4, 5 | 549 | |||
| XM_008763736.1 | XP_008761958.1 | 4, 5 | 548 | |||
| GenBank mRNA | BC097407.1 | - | 4, 5 | 506 | 7454038 | |

Hsdl2
Genomic Map