siRNA Details
| Gene Symbol: CPZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014447.2 | NP_001014447.1 | 8, 9 | 1546 | ||
| NM_001014448.2 | NP_001014448.1 | 8, 9 | 1809 | |||
| NM_003652.3 | NP_003643.2 | 7, 8 | 1513 | |||
| GenBank mRNA | U83411.1 | - | 7, 8 | 1378 | ||

CPZ
Genomic Map