siRNA Details
| Gene Symbol: Murc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107931.2 | NP_001101401.1 | 1 | 332 | ||
| GenBank mRNA | EU487255.1 | - | 1 | 332 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Murc| Gene Symbol: Murc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107931.2 | NP_001101401.1 | 1 | 332 | ||
| GenBank mRNA | EU487255.1 | - | 1 | 332 | ||