siRNA Details
| Gene Symbol: Galnt12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_232988.8 | XP_232988.5 | 10 | 1739 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Galnt12| Gene Symbol: Galnt12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_232988.8 | XP_232988.5 | 10 | 1739 | ||