siRNA Details
| Gene Symbol: Coro2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012101.2 | NP_001012101.2 | 12 | 1730 | ||
| XM_006238058.2 | XP_006238120.1 | 11 | 3285 | |||
| GenBank mRNA | BC079209.1 | - | 12 | 1683 | 7131801 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Coro2a| Gene Symbol: Coro2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012101.2 | NP_001012101.2 | 12 | 1730 | ||
| XM_006238058.2 | XP_006238120.1 | 11 | 3285 | |||
| GenBank mRNA | BC079209.1 | - | 12 | 1683 | 7131801 | |