siRNA Details
| Gene Symbol: Exosc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107936.1 | NP_001101406.1 | 2, 3 | 490 | ||
| XM_006238067.2 | XP_006238129.1 | 2, 3 | 534 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Exosc3| Gene Symbol: Exosc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107936.1 | NP_001101406.1 | 2, 3 | 490 | ||
| XM_006238067.2 | XP_006238129.1 | 2, 3 | 534 | |||