siRNA Details
| Gene Symbol: Zfp618 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763811.1 | XP_008762033.1 | 12 | 1158 | ||
| XM_008763812.1 | XP_008762034.1 | 12 | 1155 | |||
| XM_008763813.1 | XP_008762035.1 | 12 | 1134 | |||
| XM_008763814.1 | XP_008762036.1 | 12 | 1158 | |||
| XM_008763815.1 | XP_008762037.1 | 11 | 1062 | |||
| XM_008763816.1 | XP_008762038.1 | 11 | 1038 | |||

Zfp618
Genomic Map