siRNA Details
Gene Symbol: Dfnb31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181088.1 | NP_851602.1 | 12 | 2492 | ||
GenBank mRNA | AY227205.1 | - | 12 | 2492 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dfnb31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181088.1 | NP_851602.1 | 12 | 2492 | ||
GenBank mRNA | AY227205.1 | - | 12 | 2492 |