siRNA Details
| Gene Symbol: Ptprd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008763819.1 | XP_008762041.1 | 7 | 1654 | ||
| XM_008763820.1 | XP_008762042.1 | 7 | 1654 | |||
| XM_008763821.1 | XP_008762043.1 | 7 | 1654 | |||
| XM_008763822.1 | XP_008762044.1 | 6 | 1645 | |||
| XM_008763823.1 | XP_008762045.1 | 7 | 1653 | |||
| XM_008763824.1 | XP_008762046.1 | 6 | 1641 | |||
| XM_008763825.1 | XP_008762047.1 | 6 | 1636 | |||
| XM_008763826.1 | XP_008762048.1 | 5 | 1626 | |||
| XM_008763827.1 | XP_008762049.1 | 7 | 1653 | |||
| XM_008763828.1 | XP_008762050.1 | 7 | 1653 | |||
| XM_008763829.1 | XP_008762051.1 | 7 | 1653 | |||
| XM_008763830.1 | XP_008762052.1 | 7 | 1652 | |||
| XM_008763831.1 | XP_008762053.1 | 7 | 1652 | |||

Ptprd
Genomic Map