siRNA Details
| Gene Symbol: Psip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175765.2 | NP_786941.1 | 3 | 313 | ||
| XM_006238353.2 | XP_006238415.1 | 4 | 537 | |||
| XM_006238354.2 | XP_006238416.1 | 4 | 536 | |||
| XM_008763779.1 | XP_008762001.1 | 4 | 536 | |||
| GenBank mRNA | AB285525.1 | - | 216 | |||
| AF339084.1 | - | 3 | 313 | |||
| AF372090.1 | - | 3 | 313 | |||
| BC093606.1 | - | 4 | 471 | 7317952 | ||

Psip1
Genomic Map