siRNA Details
Gene Symbol: Cntln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114402.1 | NP_001107874.1 | 11, 12 | 1984 | ||
GenBank mRNA | AB369315.1 | - | 11, 12 | 1984 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cntln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114402.1 | NP_001107874.1 | 11, 12 | 1984 | ||
GenBank mRNA | AB369315.1 | - | 11, 12 | 1984 |