siRNA Details
| Gene Symbol: Focad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006238379.2 | XP_006238441.1 | 6 | 1091 | ||
| XM_233144.7 | XP_233144.5 | 12 | 1616 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Focad| Gene Symbol: Focad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006238379.2 | XP_006238441.1 | 6 | 1091 | ||
| XM_233144.7 | XP_233144.5 | 12 | 1616 | |||