siRNA Details
| Gene Symbol: Focad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006238379.2 | XP_006238441.1 | 7 | 1205 | ||
| XM_233144.7 | XP_233144.5 | 13 | 1730 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Focad| Gene Symbol: Focad | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006238379.2 | XP_006238441.1 | 7 | 1205 | ||
| XM_233144.7 | XP_233144.5 | 13 | 1730 | |||