siRNA Details
| Gene Symbol: Ift74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007001.1 | NP_001007002.1 | 15, 16 | 1349 | ||
| XM_006238398.2 | XP_006238460.1 | 15, 16 | 1457 | |||
| GenBank mRNA | BC083611.1 | - | 15, 16 | 1349 | 7133514 | |

Ift74
Genomic Map