siRNA Details
| Gene Symbol: Cyp2j3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175766.3 | NP_786942.1 | 575 | |||
| GenBank mRNA | BC089766.1 | - | 4 | 580 | 7368342 | |
| U39943.1 | - | 4 | 575 | |||
| U40000.1 | - | 589 | ||||
| U40004.1 | - | 578 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp2j3| Gene Symbol: Cyp2j3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175766.3 | NP_786942.1 | 575 | |||
| GenBank mRNA | BC089766.1 | - | 4 | 580 | 7368342 | |
| U39943.1 | - | 4 | 575 | |||
| U40000.1 | - | 589 | ||||
| U40004.1 | - | 578 | ||||