siRNA Details
| Gene Symbol: Dnajc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006238446.2 | XP_006238508.1 | 7 | 917 | ||
| XM_006238447.2 | XP_006238509.1 | 7 | 821 | |||
| XM_006238448.2 | XP_006238510.1 | 7 | 1981 | |||
| XM_006238449.2 | XP_006238511.1 | 8 | 2044 | |||

Dnajc6
Genomic Map