siRNA Details
| Gene Symbol: Wdr78 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024786.1 | NP_001019957.1 | 3 | 447 | ||
| XM_008763872.1 | XP_008762094.1 | 3 | 442 | |||
| XM_008763873.1 | XP_008762095.1 | 3 | 442 | |||
| GenBank mRNA | BC083631.1 | - | 3 | 427 | ||
| BC097402.1 | - | 3 | 447 | 7453940 | ||

Wdr78
Genomic Map