siRNA Details
Gene Symbol: Usp24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271206.1 | NP_001258135.1 | 12 | 1614 | ||
XM_006238511.2 | XP_006238573.1 | 12 | 1411 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Usp24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271206.1 | NP_001258135.1 | 12 | 1614 | ||
XM_006238511.2 | XP_006238573.1 | 12 | 1411 |