siRNA Details
| Gene Symbol: RGD1564541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107950.1 | NP_001101420.1 | 3 | 457 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1564541| Gene Symbol: RGD1564541 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107950.1 | NP_001101420.1 | 3 | 457 | ||