siRNA Details
Gene Symbol: Dock11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191760.1 | NP_001178689.1 | 31 | 3548 | ||
XM_006257453.2 | XP_006257515.1 | 32 | 3790 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dock11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191760.1 | NP_001178689.1 | 31 | 3548 | ||
XM_006257453.2 | XP_006257515.1 | 32 | 3790 |