siRNA Details
| Gene Symbol: Klhl13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002730180.4 | XP_002730226.1 | 7 | 2174 | ||
| XM_003752130.3 | XP_003752178.1 | 8 | 3954 | |||
| XM_006257443.2 | XP_006257505.1 | 10 | 2094 | |||
| XM_006257444.2 | XP_006257506.1 | 7 | 2072 | |||
| XM_006257445.2 | XP_006257507.1 | 9 | 2028 | |||
| GenBank mRNA | BC104685.1 | - | 3 | 899 | ||
| FQ228246.1 | - | 10 | 2056 | |||

Klhl13
Genomic Map