siRNA Details
| Gene Symbol: Klhl13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_002730180.4 | XP_002730226.1 | 6 | 1805 | ||
| XM_003752130.3 | XP_003752178.1 | 7 | 3585 | |||
| XM_006257443.2 | XP_006257505.1 | 9 | 1725 | |||
| XM_006257444.2 | XP_006257506.1 | 6 | 1703 | |||
| XM_006257445.2 | XP_006257507.1 | 8 | 1659 | |||
| GenBank mRNA | BC104685.1 | - | 2 | 530 | ||
| FQ228246.1 | - | 9 | 1687 | |||

Klhl13
Genomic Map