siRNA Details
| Gene Symbol: Upf3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135873.1 | NP_001129345.1 | 4, 5 | 575 | ||
| XM_006257455.2 | XP_006257517.1 | 4, 5 | 598 | |||
| XM_008773416.1 | XP_008771638.1 | 4, 5 | 598 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Upf3b| Gene Symbol: Upf3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135873.1 | NP_001129345.1 | 4, 5 | 575 | ||
| XM_006257455.2 | XP_006257517.1 | 4, 5 | 598 | |||
| XM_008773416.1 | XP_008771638.1 | 4, 5 | 598 | |||