siRNA Details
| Gene Symbol: Rnf113a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014791.1 | NP_001014791.1 | 856 | |||
| GenBank mRNA | AY363108.1 | - | 860 | |||
| BC087595.1 | - | 856 | 7310345 | |||
| Gene Symbol: Rnf113a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008773490.1 | XP_008771712.1 | 1 | 878 | ||

Rnf113a1
Genomic Map