siRNA Details
| Gene Symbol: API5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142930.1 | NP_001136402.1 | 2 | 314 | ||
| NM_001243747.1 | NP_001230676.1 | 2 | 314 | |||
| NM_006595.3 | NP_006586.1 | 2 | 314 | |||
| NR_024625.1 | 3 | 376 | ||||
| XM_006718359.3 | XP_006718422.1 | 2 | 304 | |||
| XM_017018464.1 | XP_016873953.1 | 3 | 361 | |||
| XM_017018465.1 | XP_016873954.1 | 3 | 361 | |||
| GenBank mRNA | AF229254.1 | - | 2 | 254 | ||
| AK293303.1 | - | 2 | 215 | |||
| AK299288.1 | - | 2 | 280 | |||
| AK304157.1 | - | 3 | 342 | |||
| AY311389.1 | - | 2 | 253 | |||
| BC017709.1 | - | 2 | 234 | 4419401 | ||
| BM193468.1 | - | 2 | 296 | |||
| BT007093.1 | - | 2 | 141 | |||
| DA104687.1 | - | 2 | 315 | |||
| DC334531.1 | - | 2 | 314 | |||
| U83857.1 | - | 2 | 236 | |||
| Y15906.1 | - | 2 | 273 | |||

API5
Genomic Map