siRNA Details
Gene Symbol: Eps15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009424.1 | NP_001009424.1 | 15 | 1592 | ||
GenBank mRNA | AB262963.1 | - | 15 | 1448 | ||
AY190609.1 | - | 15 | 1592 | |||
FQ223380.1 | - | 1580 |
