siRNA Details
| Gene Symbol: Orc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177931.2 | NP_808792.1 | 12 | 1951 | ||
| XM_006238515.2 | XP_006238577.1 | 13 | 2153 | |||
| XM_006238516.2 | XP_006238578.1 | 14 | 2266 | |||
| XM_006238518.2 | XP_006238580.1 | 13 | 2153 | |||
| GenBank mRNA | AB105378.1 | - | 12 | 1951 | ||

Orc1
Genomic Map