siRNA Details
| Gene Symbol: Tln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039025.2 | NP_001034114.2 | 11 | 1715 | ||
| XM_008763674.1 | XP_008761896.1 | 11 | 1483 | |||
| XM_008763675.1 | XP_008761897.1 | 12 | 3197 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tln1| Gene Symbol: Tln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039025.2 | NP_001034114.2 | 11 | 1715 | ||
| XM_008763674.1 | XP_008761896.1 | 11 | 1483 | |||
| XM_008763675.1 | XP_008761897.1 | 12 | 3197 | |||