siRNA Details
| Gene Symbol: Ccdc107 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749929.3 | XP_003749977.1 | 5 | 543 | ||
| XM_006238135.2 | XP_006238197.1 | 5 | 555 | |||
| XM_006238136.2 | XP_006238198.1 | 5 | 551 | |||
| XM_006238137.2 | XP_006238199.1 | 5 | 547 | |||
| GenBank mRNA | FQ229548.1 | - | 6 | 552 | ||

Ccdc107
Genomic Map