siRNA Details
Gene Symbol: Cd72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015016.1 | NP_001015016.1 | 1 | 134 | ||
GenBank mRNA | BC091196.1 | - | 1 | 134 | 7374499 | |
FQ233803.1 | - | 1 | 146 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cd72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015016.1 | NP_001015016.1 | 1 | 134 | ||
GenBank mRNA | BC091196.1 | - | 1 | 134 | 7374499 | |
FQ233803.1 | - | 1 | 146 |