siRNA Details
| Gene Symbol: Tal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107958.2 | NP_001101428.2 | 5 | 985 | ||
| XM_006238615.2 | XP_006238677.1 | 5 | 851 | |||
| XM_006238617.1 | XP_006238679.1 | 5 | 1059 | |||
| XM_006238618.2 | XP_006238680.1 | 4 | 703 | |||
| XM_008763967.1 | XP_008762189.1 | 5 | 1370 | |||

Tal1
Genomic Map