siRNA Details
| Gene Symbol: Atpaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107959.1 | NP_001101429.1 | 9 | 1090 | ||
| XM_006238656.2 | XP_006238718.1 | 9 | 1906 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atpaf1| Gene Symbol: Atpaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107959.1 | NP_001101429.1 | 9 | 1090 | ||
| XM_006238656.2 | XP_006238718.1 | 9 | 1906 | |||