siRNA Details
| Gene Symbol: Mob3c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107960.1 | NP_001101430.1 | 3 | 690 | ||
| XM_006238657.2 | XP_006238719.1 | 2 | 1606 | |||
| XM_006238658.2 | XP_006238720.1 | 3 | 1111 | |||
| XM_006238659.2 | XP_006238721.1 | 3 | 2608 | |||
| XM_008763977.1 | XP_008762199.1 | 2 | 1606 | |||

Mob3c
Genomic Map