siRNA Details
| Gene Symbol: Zswim5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107963.1 | NP_001101433.1 | 14 | 2303 | ||
| XM_008764009.1 | XP_008762231.1 | 14 | 2225 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zswim5| Gene Symbol: Zswim5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107963.1 | NP_001101433.1 | 14 | 2303 | ||
| XM_008764009.1 | XP_008762231.1 | 14 | 2225 | |||