siRNA Details
| Gene Symbol: Tmem53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107964.1 | NP_001101434.1 | 3 | 865 | ||
| XM_006238665.2 | XP_006238727.1 | 3 | 1455 | |||
| XM_006238666.2 | XP_006238728.1 | 2 | 678 | |||
| GenBank mRNA | FQ219156.1 | - | 3 | 819 | ||

Tmem53
Genomic Map