siRNA Details
| Gene Symbol: Olr856 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001000515.1 | NP_001000515.1 | 1 | 393 | ||
| Gene Symbol: LOC100912421 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003750008.1 | XP_003750056.1 | 1 | 393 | ||

Olr856
Genomic Map