siRNA Details
| Gene Symbol: Zfp691 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107968.2 | NP_001101438.1 | 1 | 617 | ||
| XM_006238757.2 | XP_006238819.1 | 2 | 1835 | |||
| XM_008764010.1 | XP_008762232.1 | 2 | 664 | |||
| XM_008764011.1 | XP_008762233.1 | 2 | 1116 | |||
| GenBank mRNA | BC166565.1 | - | 1 | 617 | 7316740 | |

Zfp691
Genomic Map