siRNA Details
| Gene Symbol: RGD1564804 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107969.1 | NP_001101439.1 | 4 | 409 | ||
| GenBank mRNA | FQ209773.1 | - | 4 | 435 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1564804| Gene Symbol: RGD1564804 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107969.1 | NP_001101439.1 | 4 | 409 | ||
| GenBank mRNA | FQ209773.1 | - | 4 | 435 | ||