siRNA Details
| Gene Symbol: Zmynd12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107970.1 | NP_001101440.1 | 7 | 1006 | ||
| XM_006238759.2 | XP_006238821.1 | 7 | 1021 | |||
| XM_006238760.2 | XP_006238822.1 | 7 | 1019 | |||
| XM_006238761.2 | XP_006238823.1 | 6 | 899 | |||

Zmynd12
Genomic Map