siRNA Details
| Gene Symbol: Hivep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107972.1 | NP_001101442.1 | 2 | 2520 | ||
| XM_006238779.2 | XP_006238841.1 | 3 | 5672 | |||
| XM_008764014.1 | XP_008762236.1 | 2 | 5769 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hivep3| Gene Symbol: Hivep3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107972.1 | NP_001101442.1 | 2 | 2520 | ||
| XM_006238779.2 | XP_006238841.1 | 3 | 5672 | |||
| XM_008764014.1 | XP_008762236.1 | 2 | 5769 | |||