siRNA Details
| Gene Symbol: Utp11l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107978.2 | NP_001101448.1 | 6 | 562 | ||
| GenBank mRNA | BC168964.1 | - | 6 | 562 | 7130611 | |
| FQ226285.1 | - | 6 | 574 | |||
| FQ231750.1 | - | 6 | 583 | |||
| FQ233930.1 | - | 6 | 574 | |||

Utp11l
Genomic Map