siRNA Details
| Gene Symbol: Fhl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107979.1 | NP_001101449.1 | 6 | 886 | ||
| XM_008764016.1 | XP_008762238.1 | 5 | 1105 | |||
| XM_008764017.1 | XP_008762239.1 | 5 | 1032 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fhl3| Gene Symbol: Fhl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107979.1 | NP_001101449.1 | 6 | 886 | ||
| XM_008764016.1 | XP_008762238.1 | 5 | 1105 | |||
| XM_008764017.1 | XP_008762239.1 | 5 | 1032 | |||