siRNA Details
| Gene Symbol: Sf3a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025698.1 | NP_001020869.1 | 13 | 1184 | ||
| GenBank mRNA | BC099183.1 | - | 13 | 1184 | 7383264 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sf3a3| Gene Symbol: Sf3a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025698.1 | NP_001020869.1 | 13 | 1184 | ||
| GenBank mRNA | BC099183.1 | - | 13 | 1184 | 7383264 | |