siRNA Details
| Gene Symbol: Rspo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107980.1 | NP_001101450.1 | 4, 5 | 797 | ||
| XM_008764019.1 | XP_008762241.1 | 4, 5 | 910 | |||
| XM_008764020.1 | XP_008762242.1 | 4, 5 | 963 | |||
| GenBank mRNA | BC168963.1 | - | 4, 5 | 904 | ||

Rspo1
Genomic Map