siRNA Details
| Gene Symbol: Thrap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009693.1 | NP_001009693.1 | 9 | 2341 | ||
| XM_006238866.2 | XP_006238928.1 | 9 | 2321 | |||
| XM_006238867.1 | XP_006238929.1 | 9 | 1882 | |||
| XM_008764021.1 | XP_008762243.1 | 10 | 2440 | |||
| XM_008764022.1 | XP_008762244.1 | 9 | 2690 | |||
| GenBank mRNA | BC088415.1 | - | 9 | 2341 | 7110766 | |

Thrap3
Genomic Map