siRNA Details
| Gene Symbol: Zmym1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107983.1 | NP_001101453.1 | 8 | 2689 | ||
| XM_006238878.2 | XP_006238940.1 | 6 | 3893 | |||
| XM_006238879.1 | XP_006238941.1 | 8 | 2603 | |||
| XM_006238880.2 | XP_006238942.1 | 7 | 2627 | |||
| GenBank mRNA | BC160845.1 | - | 7 | 2582 | ||
| FQ219503.1 | - | 1 | 338 | |||

Zmym1
Genomic Map