siRNA Details
| Gene Symbol: Pafah2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177932.2 | NP_808793.1 | 3 | 317 | ||
| XM_006239127.2 | XP_006239189.1 | 4 | 618 | |||
| XM_008764144.1 | XP_008762366.1 | 4 | 620 | |||
| GenBank mRNA | AY225592.1 | - | 3 | 317 | ||

Pafah2
Genomic Map